
2,5-Dithiobiurea ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:142-46-1
:
Formula:C₂H₆N₄S₂
MDL Number:MFCD00014471
:150.22 g/mol
EINECS:205-537-6
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):35.50 to 38.20 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together