

N,N-Diethyl-N'-methylethylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:104-79-0
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00009053
:130.24 g/mol
Boiling Pt:156°C
Flash Pt:41°C
EINECS:203-238-5
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together