Order Entry
Italy
ContactUsLinkComponent
3,3-Dimethyl-1-hexene ≥98.0% (by GC)
Flammable
Health
3,3-Dimethyl-1-hexene ≥98.0% (by GC)
Catalog # TCIAD1243-1ML
Supplier:  TCI
CAS Number:  
Flammable
Health
3,3-Dimethyl-1-hexene ≥98.0% (by GC)
Catalog # TCIAD1243-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :112.22 g/mol
Boiling Pt:104°C
UN:3295
ADR:3,II
 :GHS02
 :GHS08
CAS Number:3404-77-1
Formula:C₈H₁₆
MDL Number:MFCD00048727