
3,6-Dimethylphenanthrene ≥97.0% (by GC)
Catalog # TCIAD1235-100MG
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
About this item
:
:206.29 g/mol
EINECS:216-409-4
:GHS07
CAS Number:1576-67-6
Formula:C₁₆H₁₄
MDL Number:MFCD00001170