Order Entry
Italy
ContactUsLinkComponent
 
 
2,9-Dimethyl-5-decyne ≥97.0% (by GC)
  TCIAD1193-5ML
 :  TCI
 :  
 
2,9-Dimethyl-5-decyne ≥97.0% (by GC)
  TCIAD1193-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :166.31 g/mol
 :92°C
 :GHS08
 :19550-56-2
 :C₁₂H₂₂
 :MFCD00041611