
3,6-Dimethyl-4-octyne-3,6-diol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:78-66-0
:
Formula:C₁₀H₁₈O₂
MDL Number:MFCD00004480
:170.25 g/mol
EINECS:201-131-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 55.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together