Order Entry
Italy
ContactUsLinkComponent
4,4'-Dipropoxyazoxybenzene ≥98.0% (by total nitrogen basis)
4,4'-Dipropoxyazoxybenzene ≥98.0% (by total nitrogen basis)
Catalog # TCIAD1094-5G
Supplier:  TCI
CAS Number:  
4,4'-Dipropoxyazoxybenzene ≥98.0% (by total nitrogen basis)
Catalog # TCIAD1094-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Mesomorphic range:
    115 to 128 deg-C

Specification Test Results

About this item

 :
 :314.39 g/mol
CAS Number:23315-55-1
Formula:C₁₈H₂₂N₂O₃
MDL Number:MFCD00059870