4,4'-Diamyloxyazoxybenzene ≥95.0% (by total nitrogen basis)
Catalog # TCIAD1093-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (with Total Nitrogen):min. 95.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:370.49 g/mol
CAS Number:19482-05-4
Formula:C₂₂H₃₀N₂O₃
MDL Number:MFCD00041933