4,4'-Dihexyloxybiphenyl ≥98.0% (by GC)
Catalog # TCIAD1089-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:127.0 to 130.0 deg-C
Specification Test Results
About this item
:
:354.53 g/mol
CAS Number:142450-58-6
Formula:C₂₄H₃₄O₂
MDL Number:MFCD00059438