4,4'-Dibutoxybiphenyl ≥98.0% (by GC)
Catalog # TCIAD1082-25G
Supplier: TCI
CAS Number:
Specifications
- Conf:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in THF:almost transparency
Specification Test Results
About this item
:
:298.43 g/mol
CAS Number:39800-63-0
Formula:C₂₀H₂₆O₂
MDL Number:MFCD00059419