


Sodium dodecylbenzenesulphonate mixture ≥95.0% (by titrimetric analysis), hard type
Supplier: TCI
Certificates
About this item
CAS Number:69227-09-4
:
Formula:C₁₈H₂₉NaO₃S
MDL Number:MFCD00011508
:348.48 g/mol
Merck Index:14,08612
EINECS:246-680-4
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Gravimetric Method):95.0 to 105.0 % (calcd.on dried substance)
- Drying loss:max. 5.0 %
- pH:5.5 to 8.0 (10g/L, 25 deg-C)
- Petroleum ether soluble matter:max. 2.0 %
Specification Test Results
Frequently Bought Together