
1,3-Di(4-pyridyl)propane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17252-51-6
:
Formula:C₁₃H₁₄N₂
MDL Number:MFCD00038046
:198.27 g/mol
Boiling Pt:162°C
EINECS:241-284-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:60.0 to 64.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together