
Diisopropylamine hydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:819-79-4
:
Formula:C₆H₁₅N
MDL Number:MFCD00034860
:137.65 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:215.0 to 220.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together