
Di-isopropanolamine mixture of DL/meso ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:110-97-4
:
Formula:C₆H₁₅NO₂
MDL Number:MFCD00004531
:133.19 g/mol
Merck Index:14,03195
EINECS:203-820-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 90.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together