Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Di-tert-octyl disulphide
Di-tert-octyl disulphide ≥75.0% (by GC)
Di-tert-octyl disulphide ≥75.0% (by GC)
TCIAD0858-25ML
:
TCI
:
Di-tert-octyl disulphide ≥75.0% (by GC)
TCIAD0858-25ML
:
TCI
:
Pk:
25 mL
Seq:
2315
Appearance:
White to Yellow to Green clear liquid
Purity (GC):
min. 75.0 %
:
:
290.57 g/mol
:
249-974-0
:
29956-99-8
:
C₁₆H₃₄S₂
:
MFCD00048262