
3,5-Dinitrosalicylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:609-99-4
:
Formula:C₇H₄N₂O₇
MDL Number:MFCD00007104
:228.12 g/mol
EINECS:210-204-3
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:172.0 to 175.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together