
3,3'-Dinitrobenzidine ≥95.0% (by HPLC)
Catalog # TCIAD0822-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
About this item
:
:274.24 g/mol
Melting Pt:275°C
EINECS:228-457-3
:GHS07
CAS Number:6271-79-0
Formula:C₁₂H₁₀N₄O₄
MDL Number:MFCD00024264