
N,N-Dimethyl-1,4-phenylenediamineoxalate ≥98.0% (by titrimetric analysis)
Codice catalogo TCIAD0781-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
ADR:6.1,II
:362.43 g/mol
EINECS:263-723-2
UN:2811
:GHS06
CAS Number:62778-12-5
Formula:C₈H₁₂N₂
MDL Number:MFCD00042038