Order Entry
Italy
ContactUsLinkComponent
 
 
3,3'-Dimethylnaphthidine ≥97.0% (by titrimetric analysis) for colorimetric determination of Cl in water
 :  TCI
 
 
3,3'-Dimethylnaphthidine ≥97.0% (by titrimetric analysis) for colorimetric determination of Cl in water
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Sensitiveness:
    Colorimetric Test : Turn Yellow with 0.1ppm Chloride ion

 

Frequently Bought Together