

Arsenazo I hydrate ≥90.0% (by titrimetric analysis)
Catalog # TCIAD0578-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Dark red to Brown powder to crystal
- Purity (Iodometric Titration):min. 90.0% (calcd.on anh.substance)
- Absorbance:min. 0.700 (20 mg/L, pH 6.3, 498.0 to 502.0 nm)
- Drying loss:max. 5.0 %
- Ignition residue (Sulfate):32.0 to 37.0 % (calcd.on anh.substance)
Specification Test Results
About this item
:
:614.27 g/mol
EINECS:266-069-6
UN:3465
ADR:6.1,III
:GHS06
:GHS09
CAS Number:3547-38-4
Formula:C₁₆H₁₃AsN₂O₁₁S₂·xH₂O
MDL Number:MFCD00066725