
Digitoxigenin ≥97.0% (by HPLC)
Catalog # TCIAD0541-10MG
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:+15.0 to +20.0 deg (C=1, MeOH)
- Lambda max.:218.0 to 222.0 nm (EtOH (50%))
- Molar extinction coefficient:14000 to 18000 (EtOH (50%), 218.0 to 222.0 nm)
Specification Test Results
About this item
:
:374.52 g/mol
Merck Index:14,03162
Melting Pt:253°C
EINECS:205-603-4
UN:2811
ADR:6.1,II
:GHS06
CAS Number:143-62-4
Formula:C₂₃H₃₄O₄
MDL Number:MFCD00003687