Order Entry
Italy
ContactUsLinkComponent
N,N-Diethyl-1-naphthylamine ≥98.0% (by GC)
N,N-Diethyl-1-naphthylamine ≥98.0% (by GC)
Catalog # TCIAD0513-10G
Supplier:  TCI
CAS Number:  
N,N-Diethyl-1-naphthylamine ≥98.0% (by GC)
Catalog # TCIAD0513-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :199.30 g/mol
Boiling Pt:285°C
EINECS:201-576-8
CAS Number:84-95-7
Formula:C₁₄H₁₇N
MDL Number:MFCD00046389