Order Entry
Italy
ContactUsLinkComponent
1,3-Di-4-piperidylpropane ≥97.0% (by GC)
Irritant
1,3-Di-4-piperidylpropane ≥97.0% (by GC)
Catalog # TCIAD0293-25G
Supplier:  TCI
CAS Number:  
Irritant
1,3-Di-4-piperidylpropane ≥97.0% (by GC)
Catalog # TCIAD0293-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    65.0 to 68.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :210.37 g/mol
EINECS:240-941-6
 :GHS07
CAS Number:16898-52-5
Formula:C₁₃H₂₆N₂
MDL Number:MFCD00038013