4,4'-Dithiodimorpholine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:103-34-4
:
Formula:C₈H₁₆N₂O₂S₂
MDL Number:MFCD00023319
Merck Index:14,03372
Melting Pt:125°C
:236.35 g/mol
Storage Temperature:Refrigerator
EINECS:203-103-0
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together