Order Entry
Italy
ContactUsLinkComponent
1,3-Diisopropylthiourea ≥99.0% (by total nitrogen basis)
Irritant
1,3-Diisopropylthiourea ≥99.0% (by total nitrogen basis)
Catalog # TCIAD0253-25G
Supplier:  TCI
CAS Number:  
Irritant
1,3-Diisopropylthiourea ≥99.0% (by total nitrogen basis)
Catalog # TCIAD0253-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 99.0 %
  • Melting point:
    143.0 to 146.0 deg-C

Specification Test Results

About this item

 :
 :160.28 g/mol
EINECS:221-051-7
 :GHS07
CAS Number:2986-17-6
Formula:C₇H₁₆N₂S
MDL Number:MFCD00040485