4,4'-Dibenzoylquinone dioxime ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:120-52-5
:
Formula:C₂₀H₁₄N₂O₄
MDL Number:MFCD00045557
:346.34 g/mol
EINECS:204-403-4
Specification Test Results
- Appearance:Light gray to Brown to Black powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together