
4-Diazo-N,N-dimethylaniline chloride zinc chloride hydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6087-56-5
:
Formula:C₈H₁₀ClN₃
MDL Number:MFCD00031685
:319.92 g/mol
EINECS:228-021-2
UN:1325
ADR:4.1,II
:GHS02
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Argentmetric Titration):min. 95.0 % (calcd.on dried substance)
- Purity (Chelometric Titration):min. 95.0 % (calcd.on dried substance)
- Drying loss:max. 20.0 %
Specification Test Results
Frequently Bought Together