
1,5-Diaminopentane dihydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1476-39-7
:
Formula:NH₂(CH₂)₅NH₂·2HCl
MDL Number:MFCD00012527
:175.10 g/mol
EINECS:216-022-0
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together