3,20-Dioxopregn-4-en-21-yl acetate ≥98.0% (by HPLC)
Catalog # TCIAD0047-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:156.0 to 162.0 deg-C
- Specific rotation [a]20/D:+170.0 to +180.0 deg (C=1, Dioxane)
- Solubility in Dioxane:almost transparency
Specification Test Results
About this item
:
:372.51 g/mol
Merck Index:14,02901
EINECS:200-275-9
CAS Number:56-47-3
Formula:C₂₃H₃₂O₄
MDL Number:MFCD00003660