
4-Chloro-3-ethyl-1-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:127892-62-0
:
Formula:C₇H₉ClN₂O₂
MDL Number:MFCD08459057
:188.61 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
Frequently Bought Together