
2-Cyano-2-propyl Dodecyl trithiocarbonate ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAC3627-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Specific gravity (20/20):0.9950 to 1.0050
Specification Test Results
About this item
:
:345.62 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:870196-83-1
Formula:C₁₇H₃₁NS₃
MDL Number:MFCD11973800