Cholesterol ≥99.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:57-88-5
:
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646
:386.66 g/mol
Merck Index:14,02201
:Stabilised with α-tocopherol
Storage Temperature:Freezer
EINECS:200-353-2
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:148.0 to 151.0 deg-C
- Specific rotation [a]20/D:-34.0 to -39.5 deg (C=2, Dioxane)
- Drying loss:max. 0.2 %
Specification Test Results
Frequently Bought Together