
Cobalt(II) meso-tetraphenylporphine ≥80.0% (by HPLC)
Catalog # TCIAC3579-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Purity (HPLC):min. 80.0 area%
- Purity (Chelometric Titration):min. 95.0 %
- Molar extinction coefficient:min. 160000 (THF, 415.0 to 425.0nm)
Specification Test Results
About this item
:
:671.65 g/mol
EINECS:238-018-8
:GHS08
CAS Number:14172-90-8
Formula:C₄₄H₂₈CoN₄
MDL Number:MFCD00010725