SC-560 ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:188817-13-2
:
Formula:C₁₇H₁₂N₂OClF₃
MDL Number:MFCD02179214
:352.7 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):7.50 to 8.50 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
Frequently Bought Together