Order Entry
Italy
ContactUsLinkComponent
 
Cycloheptane-1,3-dione ≥95.0% (by GC, titration analysis)
  TCIAC3564-1G
 :  TCI
 :  
Cycloheptane-1,3-dione ≥95.0% (by GC, titration analysis)
  TCIAC3564-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

 

 

 :
 :126.155 g/mol
 :122°C
 :1194-18-9
 :C₇H₁₀O₂
 :MFCD01863735