Order Entry
Italy
ContactUsLinkComponent
 
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥98.0% (by HPLC)
 :  TCI
 
undefined
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 % (calcd.on anh.substance)
  • Water:
    max. 6.0 %

 

Frequently Bought Together