Order Entry
Italy
ContactUsLinkComponent
6-Chloropyrimidine-4,5-diamine ≥98.0% (by GC, titration analysis)
Irritant
6-Chloropyrimidine-4,5-diamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC3433-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
6-Chloropyrimidine-4,5-diamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC3433-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :144.5600 g/mol
EINECS:224-341-1
 :GHS07
CAS Number:4316-98-7
Formula:C₄H₅ClN₄
MDL Number:MFCD00023270