
3-Chloro-5-fluorophenylacetonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:493038-93-0
:
Formula:C₈H₅ClFN
MDL Number:MFCD04038259
:169.59 g/mol
UN:3276
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together