
4-Chloro-2-(trifluoromethyl)quinoline ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1701-24-2
:
Formula:C₁₀H₅ClF₃N
MDL Number:MFCD00153105
:231.61 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:34.0 to 38.0 deg-C
Specification Test Results
Frequently Bought Together