
Cobalt(II) Phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Catalog # TCIAC3252-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Green to Dark green to Dark blue powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Molar extinction coefficient:min. 110000 (Pyridine, 654.0 to 659.0nm)
- Molar extinction coefficient2:min. 30000 (Pyridine, 594.0 to 599.0nm)
Specification Test Results
About this item
:
:571.47 g/mol
Melting Pt:300°C
EINECS:222-012-7
:GHS08
CAS Number:3317-67-7
Formula:C₃₂H₁₆CoN₈
MDL Number:MFCD00010718