Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:286961-15-7
:
Formula:C₁₉H₂₆BNO₄
MDL Number:MFCD11521562
:343.23 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
Specification Test Results
Frequently Bought Together