Order Entry
Italy
ContactUsLinkComponent
Chloro(1,5-hexadiene)rhodium(I) dimer ≥93.0% (by titrimetric analysis)
Irritant
Chloro(1,5-hexadiene)rhodium(I) dimer ≥93.0% (by titrimetric analysis)
Catalog # TCIAC3194-100MG
Supplier:  TCI
CAS Number:  
Irritant
Chloro(1,5-hexadiene)rhodium(I) dimer ≥93.0% (by titrimetric analysis)
Catalog # TCIAC3194-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (Argentmetric Titration):
    min. 93.0 %

Specification Test Results

About this item

 :
 :434.96 g/mol
Melting Pt:120°C
 :GHS07
CAS Number:32965-49-4
Formula:C₁₂H₂₀Cl₂Rh₂
MDL Number:MFCD00064595