8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:31251-41-9
:
Formula:C₁₄H₁₀ClNO
MDL Number:MFCD00800222
:243.69 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
Frequently Bought Together