
4-Cyanophenyl isothiocyanate ≥98.0% (by GC)
TCIAC3169-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:119.0 to 123.0 deg-C
:
:160.2 g/mol
:Refrigerator
:220-323-2
:2811
:6.1,III
:GHS06
:2719-32-6
:C₈H₄N₂S
:MFCD00041085