
4-Chloro-3-methylphenylboronic acid (contains varying amounts of Anhydride)
: TCI
:161950-10-3
:
:C₇H₈BClO₂
:MFCD02683104
:258°C
:170.40 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Frequently Bought Together