
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine ≥96.0% (by HPLC, total nitrogen)
TCIAC3036-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:145.0 to 149.0 deg-C
:
:307.7600 g/mol
:GHS07
:479633-63-1
:C₁₃H₁₀ClN₃O₂S
:MFCD09907939