Order Entry
Italy
ContactUsLinkComponent
2-(4-Chlorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
2-(4-Chlorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAC2990-200MG
Supplier:  TCI
CAS Number:  
2-(4-Chlorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAC2990-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :330.82 g/mol
CAS Number:5496-32-2
 :MFCD09028023
Formula:C₂₁H₁₅ClN₂
MDL Number:MFCD00030329