Order Entry
Italy
ContactUsLinkComponent
 
(1R,2R)-1,2-Cyclohexanedimethanol ≥98.0% (by GC)
 :  TCI
 
(1R,2R)-1,2-Cyclohexanedimethanol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    63.0 to 67.0 deg-C
  • Specific rotation [a]20/D:
    +16.0 to +20.0 deg (C=1, toluene)

 

Frequently Bought Together