
5-Chlorothiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:42518-98-9
:
Formula:C5H2Cl2OS
MDL Number:MFCD00043886
:181.04 g/mol
Boiling Pt:225°C
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together