Order Entry
Italy
ContactUsLinkComponent
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Irritant
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Catalog # TCIAC2824-100MG
Supplier:  TCI
CAS Number:  
Irritant
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Catalog # TCIAC2824-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Elemental analysis (Nitrogen):
    1.50 to 1.80 %

Specification Test Results

About this item

 :
 :804.90 g/mol
 :GHS07
CAS Number:64536-78-3
Formula:C₃₁H₅₀F₆IrNP₂
MDL Number:MFCD00075097